PsiI-v2
Supplier: New England Biolabs (NEB)
R0744L R0744S
76339-762EA 555 USD
76339-762 76339-764
PsiI-v2
Nucleic Acid Reagents Restriction Enzymes
A restriction endonuclease that recognizes the sequence TTA^TAA.
  • 100% activity in CutSmart buffer (over 215 enzymes are available in the same buffer)
  • Supplied with 1 vial of gel loading dye, Purple (6X)