NEBNext® Q5U® Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
M0597L M0597S
76339-746EA 530 USD
76339-746 76339-748
NEBNext® Q5U® Master Mix, New England Biolabs
Nucleic Acid Reagents RT-PCR and cDNA Reagents
NEBNext® Q5U® master mix contains a modified version of Q5® High Fidelity DNA Polymerase, optimized for amplification of uracil-containing NGS libraries. This hot start formulation is designed to read through uracils, providing robust, high-fidelity and uniform library amplification.
  • Optimized for high yields in uracil-containing NGS library amplification
  • Compatible with NEBNext® Enzymatic Methyl-seq (EM-seq™) and bisulfite sequencing workflows
  • Minimizes GC bias, with superior performance across the GC spectrum
  • Ultra-high-fidelity amplification
  • Aptamer-based hot start for room-temperature reaction set-up without a separate activation step
NEBNext Q5U master mix is included as a component in the NEBNext® Enzymatic Methyl-seq (EM-seq™) kit (76337-184, 76337-038) and is also compatible with library amplification in bisulfite sequencing workflows.