SP Bel-Art® Long-Handled Dippers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36780-0016
47727-714CS 2757.04 USD
47727-714 76339-338
SP Bel-Art® Long-Handled Dippers, Bel-Art Products, a part of SP
Samplers Sample Dippers
For easy sampling from tanks, vats, lakes, etc., these high-density polyethylene plastic dippers will not rust or corrode.
  • Handles are 2.9 cm (1¹/₈") in diameter and welded to a rigid sampling bowl at a 45° angle for easier sampling from an elevated position
  • Hook at end for hanging storage
  • Hardwood dowel totally encased in 12' handles adds rigidity and strength
Dippers provide excellent chemical resistance, and will float, 'just in case'.