Oakton® CON 550 Benchtop Conductivity Meter, Cole-Parmer®
Supplier: Cole-Parmer
WD-35419-35
76338-518EA 674.88 USD
76338-518
Oakton® CON 550 Benchtop Conductivity Meter, Cole-Parmer®
Electrochemistry Meters Conductivity Meters
Economical meter for benchtop conductivity measurement.
  • Automatically recognizes calibration solutions
  • Compact design saves valuable benchtop space
  • Data logging allows you to store and recall up to 50 groups of data
  • IP54-rated housing is dust- and spill-proof
The CON 550 benchtop conductivity meter features a microprocessor chip which provides automatic calibration, automatic temperature compensation, data storage, parameter setup, and Max/Min reading display. The meter reads 0 to 200 mS/cm across four ranges and the four-point conductivity calibration, with automatic solution recognition, makes for easy calibration. The advanced digital processing technology improves meter response time and measuring accuracy. A stable reading display icon appears when the measuring value is stable.
A conductivity electrode with temperature sensor measures conductivity and temperature simultaneously and allows for automatic temperature compensation. The compact meter includes a detachable electrode holder and three bottles of conductivity calibration solution. It has a dustproof and spillproof rating of IP54. Connectors are protected by silicone seal caps.
Certifications: CE compliant.
Accessories information: 550 Series Benchtop Meter Accessories – choose from a wide range of electrodes for your application. Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Delivery information: Plastic combination conductivity electrode (K=1.0) with temperature sensor, electrode holder, conductivity standard calibration solutions (84 µS/1413 µS/12.88 mS, 50 ml each), and 9 VAC power adapter.