Lead (Test), granulated silver-free

Supplier: ALPHA ASSEMBLY SOLUTIONS, INC.

10607-20KG
76338-316EA 1343.92 USD
76338-316
Lead (Test), granulated silver-free
Lead

Used for analysis of precious metals, Test Lead (or lead shot) is typically used in both Fire Assay bullion work and cupellation.

Formula: Pb
MW: 207.2 g/mol
MDL Number: MFCD00134050
CAS Number: 7439-92-1

Order Now

Specification Test Results

Silver ≤0.4 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR