Rectangular Basket, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00-02035002-31
76338-212EA 663.54 USD
76338-212
Rectangular Basket, Marlin Steel Wire Products
Baskets Wire Baskets
Stainless steel material handling and parts washing basket with removable lid.
  • Electropolished finish
  • With four mesh openings per linear inch
Basket features rugged construction perfect for applications such as dipping, cleaning, or moving components in an industrial setting.