Ultrasonic Cleaning Basket, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00-00368050-31
76338-116EA 818.66 USD
76338-116
Ultrasonic Cleaning Basket, Marlin Steel Wire Products
Baskets Wire Baskets
Stainless steel material handling parts washing basket with a lid and hinge.
  • Electropolished finish
  • With four mesh openings per linear inch
Great for material handling and parts washing/ultrasonic cleaning.
Rugged construction ensures basket longevity in applications such as dipping, cleaning, or moving components in industrial settings.