Mesh Basket with Hinged Lid, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
01062001-38
76338-110EA 741.3 USD
76338-110
Mesh Basket with Hinged Lid, Marlin Steel Wire Products
Baskets Wire Baskets
This stainless steel basket has a hinged lid to make sure your contents are secure.
  • Electropolished finish
  • With 12 mesh openings per linear inch
It is Ideal for storage and materials handling.