Mesh Basket, Rectangular, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00-02035001-38
76338-084EA 556.61 USD
76338-084
Mesh Basket, Rectangular, Marlin Steel Wire Products
Baskets Wire Baskets
Stainless steel material handling parts washing basket with handles.
  • Natural finish
  • With four mesh openings per linear inch
  • Rugged, with additional reinforcements to allow stacking
Basket is perfect for dipping, cleaning, or moving components in industrial environments.