Mesh Baskets, Round, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00-100-31 00-101-31 00-102-31
76338-058EA 371.29 USD
76338-058 76338-060 76338-062
Mesh Baskets, Round, Marlin Steel Wire Products
Baskets Wire Baskets
Round material handling basket with handles and mesh.
  • Electropolished finish
  • With four mesh openings per linear inch
  • Handles for easy gripping
  • Stacks so that parts inside the basket do not get crushed
Robust quality that is ideal for washing parts, cleaning, degreasing, ultrasonic cleaning, curing and materials handling in a variety of applications such as chemical baths, heat treating and cryogenics.