Myco-Sniff-Valid™ Mycoplasma PCR Kit, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
093050301
76337-170EA 405.94 USD
76337-170
Myco-Sniff-Valid™ Mycoplasma PCR Kit, MP Biomedicals
Assays qPCR Assays
Myco-Sniff-Valid™ mycoplasma PCR detection it provides a highly sensitive, specific and rapid method for the detection of a broad range of mycoplasma species in cell cultures.
  • Easy to use: Contains premixed components for the PCR reaction, just add template DNA or samples
  • Highly specific: No interference of animal or bacterial DNA
  • Exogenous internal control: Helps differentiate false negatives due to PCR inhibition or erroneous PCR tests
The detection limit for contaminating mycoplasma is as low as 10 CFU/ml. The kit is sufficient for 48 tests (20 μl reactions).