Myco-Sniff™ Mycoplasma PCR Kit, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
093050201
76337-168EA 399.84 USD
76337-168
Myco-Sniff™ Mycoplasma PCR Kit, MP Biomedicals
Assays qPCR Assays
Myco-Sniff™ mycoplasma PCR detection kit provides a highly sensitive, specific and rapid method for the detection of a broad range of mycoplasma species in cell cultures.
  • Highly specific: No Interference of animal or bacterial DNA
  • Sample control: Easily verify the effectiveness of template gDNA by checking the amplification of the sample control DNA
The detection limit for contaminating mycoplasma is as low as 20 CFU/ml. Includes 48 lyophilized PCR tubes for 20 μl reactions.