FastCounting™ Cell Counting Kits, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
The FastCounting™ cell counting kits provide a sensitive and convenient method to determine cell viability in cell proliferation and cytotoxicity assays.
- Benefits of using FastCounting cell kit
- Highest sensitivity dye for cell viability
- Lowest cytotoxicity among all tetrazolium reagents
- Simple procedures without thawing reagents
- Long shelf life and easy storage
- Store at –20 °C long-term
The kit uses a highly sensitive tetrazolium dye with low cytotoxicity to quantify the number of living cells. Sufficient for 100 tests. The FastCounting cell counting kit is stable for over one year at 2 to 8 °C when protected from light.
Caution: Avoid repeated thawing and freezing, which can cause an increase in background.