pZerve™ Cryopreservation Solution, MP Biomedicals
Supplier: 0000012207
092030346 0920303M2
76337-142EA 301.91 USD
76337-142 76337-144
pZerve™ Cryopreservation Solution, MP Biomedicals
Cell Culture Supplements and Additives
pZerve™ Cyropreservation solution is a ready-to-use, sterile, cryopreservation solution lacking dimethyl sulfoxide (DMSO), fetal bovine serum or other animal proteins.
This solution has been successfully used with the following cell types including, but not limited to mast cells, dendritic cells, mesenchymal cells, LNCaP - Metastatic Prostate Adenocarcinoma, Human, LS 180 - Colon, Adenocarcinoma, Human and 293 - Transformed Primary Embryonal Kidney, Human. For use as cell culture and in cell cryopreservation.
This solution does not require any dilution or further processing and can be used for cells cultured in media with and without serum.