Penicillin : Streptomycin solution (20,000 IU Pen/mL, 20,000 µg Strep/mL), sterile

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

091670449 091670454
76337-138EA 56.78 USD
76337-138 76337-140
Penicillin : Streptomycin solution (20,000 IU Pen/mL, 20,000 µg Strep/mL), sterile
Penicillin/Streptomycin

Double antibiotic solution containing 20,000 units/mL of penicillin and 20,000 µg/mL of streptomycin.


  • Dual antibiotic solution
  • Supplements cell culture media
  • Effective against most gram positive and gram negative bacteria


Penicillin-Streptomycin is is a dual antibiotic solution used to supplement cell culture media to control bacterial contamination. The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. This solution contains 17,000 - 21,000 units/mL of penicillin and 18.0 - 23.0 mg/mL of streptomycin.


Caution: For laboratory use. Not for injection.

Storage Temperature: Freezer
CAS Number: 1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR