Sani-Cloth® Bleach Germicidal Disposable Wipes, PDI®
Supplier: Healthcare Products
PDI P54072 PDI P7007P PDI P700RF PDI H58195 PDI P25784 PDI U26595
76337-020CS 271.44 USD
76337-020 76337-022 76337-024 76337-062 76337-092 76337-108
Sani-Cloth® Bleach Germicidal Disposable Wipes, PDI®
Wipes Disposable Wipes and Towels Disinfectant Wipes
Ideal for disinfecting high risk areas endemic with multi-drug resistant organisms, Clostridium, difficile spores, and Norovirus.
  • Compatible with a broad range of surfaces and equipment in healthcare
  • Up to 2× greater compatibility vs competitors with materials used in healthcare
  • 20% extended equipment life compared to competitors
  • No visible degradation on stainless steel, ABS plastic, brass alloy, and more
Effective against 50 micro organisms in four minutes, including the following MDRO's, blood borne pathogens, and viruses: C.difficile spores, Acinetobacter baumannii, Klebsiella pneumoniae, Candida albicans, ESB-resistant E. coli, ESB-resistant Klebsiella pneumoniae, Norovirus, MRSA, VRE, VRSA, HIV, HBV, and HCV.
Wipes may be used on hard, non porous surfaces in healthcare facilities such as bed railings, blood glucose meters (glucometers), cabinets, carts, chairs, counters, dental unit instruments trays, exam tables, gurneys, isolettes, IV poles, stethoscopes, stretchers, tables, telephones, toilet seats, diagnostic equipment, patient monitoring equipment, patient support and delivery equipment, and more. Please refer to the product/master label to see a full listing of use sites.