Sani-Cloth® Prime Germicidal Disposable Wipes, PDI®
Supplier: Healthcare Products
PDI P25372
76337-090CS 194.88 USD
76337-090
Sani-Cloth® Prime Germicidal Disposable Wipes, PDI®
Wipes Disposable Wipes and Towels Disinfectant Wipes
Powered by ACCELOQUAT™ formulation, a next generation proprietary blend of quaternary ammonium, isopropyl alcohol (IPA), and ethanol. This unique combination delivers a true 1-minute bactericidal, fungicidal, virucidal, and tuberculocidal disinfectant. It also provides the power to destroy over 50 microorganisms, including 17 multi-drug resistant organisms (MDROs) like MRSA, CRE, and VRE.
  • Disinfection level: EPA-registered intermediate level disinfectant
  • Efficacy: Bactericidal, virucidal, fungicidal, tuberculocidal
Sani-Cloth® Prime Germicidal Disposable Wipes may be used on hard nonporous surfaces in healthcare facilities such as, bed railings, blood glucose meters (glucometers), cabinets, carts, chairs, counters, dental unit instruments trays, exam tables, gurneys, isolettes, IV poles, stethoscopes, stretchers, tables, telephones, toilet seats, diagnostic equipment, patient monitoring equipment, patient support and delivery equipment, and more. Please refer to the product/master label to get the full listing of use sites.