Sani-Cloth® AF3 Germicidal Disposable Wipe, PDI®
Supplier: Healthcare Products
PDI H59200 PDI M8063S80 PDI P2450P PDI U27500
76337-064CS 194.64 USD
76337-064 76337-078 76337-088 76337-110
Sani-Cloth® AF3 Germicidal Disposable Wipe, PDI®
Wipes Disposable Wipes and Towels Disinfectant Wipes
Wipes are effective against 45 microorganisms in three minutes. Protecting staff and patients has never been easier. The alcohol-free and fragrance-free formula is ideal for use around patients, residents and staff, especially those with respiratory sensitivities.
  • Short three minute contact time is effective against enveloped/non-enveloped viruses, bacteria, TB, fungi, multi-drug resistant organisms, and blood borne pathogens
  • Alcohol, fragrance, bleach, acid, phenol, acetone, and ammonia-free
  • Bactericidal, Tuberculocidal, Virucidal
Wipes may be used on hard, nonporous surfaces in healthcare facilities such as bed railings, blood glucose meters (glucometers), cabinets, carts, chairs, counters, dental unit instruments trays, exam tables, gurneys, isolettes, IV poles, stethoscopes, stretchers, tables, telephones, toilet seats, diagnostic equipment, patient monitoring equipment, patient support and delivery equipment, and more. Please refer to the product/master label to see a full listing of use sites.