Prevantics® Device Swab and Strip, PDI Healthcare
Supplier: Healthcare Products
PDI B123ST
76337-048CS 807.03 USD
76337-048
Prevantics® Device Swab and Strip, PDI Healthcare
Disinfection Systems
Disinfect needleless access sites prior to use by scrubbing the hub.
  • Confirmed efficacy at the five second scrub time and five second dry time
  • Achieved a >4.0 log10 reduction versus a leading alcohol impregnated cap against seven tested clinically relevant microorganisms known to cause CLABSIs2
The first FDA-cleared medical device featuring a 3.15% (w/v) Chlorhexidine gluconate and 70% (v/v) Isopropyl alcohol formulation.