Prevantics® Swab, Swabstick and Maxi Swabstick, PDI Healthcare
Supplier: Healthcare Products
PDI B10800
76337-046CS 390.4 USD
76337-046
Prevantics® Swab, Swabstick and Maxi Swabstick, PDI Healthcare
Disinfection Systems
Prevantics® products, designed with 3.15% Chlorhexidine Gluconate (w/v) and 70% Isopropyl Alcohol (v/v) formulation, help prevent the spread of infections and help improve patient outcomes.
  • No glass to break
  • Pre-activated
  • No waiting
  • Easy to open, easy to use
  • Pre-moistened
  • Swab: Intuitive prep-pad design
  • Swabsticks: Unique two-sided applicator for interdigital areas and skin folds
Prevantics® products are the first and only FDA approved.