pH Electrode, LE410
Supplier: Mettler Toledo
51340348
76336-674EA 375 USD
76336-674
pH Electrode, LE410
Electrodes pH Electrodes
Three in one glass pH electrode with integrated temperature sensor and with liquid electrolyte for rapid measurements in the laboratory; for aqueous samples such as acids, bases etc.
  • Affordable and Reliable
  • Ergonomic and easy
Steady companions with NTC 30 kΩ temp. probe, ceramic junction, Ag/AgCl reference system, 3 M KCl sat. AgCl electrolyte, 120 mm shaft, Ø 12 mm, 1.0 m cable, BNC.