EC 28 Series Fixed Transmitters, Fixed Systems Gas Detection, GfG Instrumentation
Supplier: GFG INSTRUMENTATION, INC.
2811-4505-002 2811-707-002 2811-709-002 2811-712-002 2811-713-002 2813-716-002 2811-736-002 2811-719-002 2811-720-002 2811-724-002 2811-722-002 2811-727-002 2811-737-002 2811-000-008 2811-4501-008 2811-4502-008 2811-4505-008 2811-731-002 2811-000-003 2811-4501-003 2811-4502-003 2811-4505-003 2811-707-003 2811-709-003 2811-712-003 2811-713-003 2811-716-003 2811-736-003 2811-719-003 2811-720-003 2811-724-003 2811-722-003 2811-727-003 2811-737-003 2811-731-003 2811-000-007 2811-4501-007 2811-4502-007 2811-4505-007 2811-707-007 2811-709-007 2811-712-007 2811-713-007 2811-716-007 2811-736-007 2811-719-007 2811-000-002 2811-4501-002 2811-4502-002 2811-720-007 2811-724-007 2811-722-007 2811-727-007 2811-737-007 2811-731-007 2811-707-008 2811-709-008 2811-712-008 2811-713-008 2811-716-008 2811-736-008 2811-719-008 2811-720-008 2811-724-008 2811-722-008 2811-727-008 2811-737-008 2811-731-008
76334-888EA 1907.28 USD
76334-888 76334-894 76334-896 76334-898 76334-900 76334-902 76334-904 76334-906 76334-908 76334-910 76334-912 76334-914 76334-916 76334-924 76334-926 76334-928 76334-930 76334-994 76334-996 76334-998 76335-000 76335-002 76335-008 76335-010 76335-012 76335-014 76335-016 76335-018 76335-020 76335-022 76335-024 76335-026 76335-028 76335-030 76335-032 76335-034 76335-036 76335-038 76335-040 76335-046 76335-048 76335-050 76335-052 76335-054 76335-056 76335-058 76335-080 76335-082 76335-084 76335-086 76335-088 76335-090 76335-092 76335-094 76335-096 76335-098 76335-100 76335-102 76335-104 76335-106 76335-108 76335-110 76335-112 76335-114 76335-116 76335-118 76335-120 76335-122
EC 28 Series Fixed Transmitters, Fixed Systems Gas Detection, GfG Instrumentation
Gas Detection Systems
Wherever hazards are expected, the EC 28 transmitter and GfG’s proven monitoring systems are the right choice for reliable surveillance. The ATEX-certified design, an integrated, Ex-proof buzzer, and bright alarm LEDs provide the highest safety in hazardous areas by including visual and audible alarms (optional). The gas alarm not only alerts the person in the control room of hazards, but also the person on-site.
  • Increased safety design to reduce installation cost in potentially hazardous locations
  • Non-intrusive AutoCal® calibration adjustment
  • Uses electrochemical detection principle
  • Output signal: 4 to 20 mA
  • Available with MODBUS
Available in seven configurations:
Blind (no display)
Display
Display with local alarms
Display with local alarms and relays
Blind (no display) with MODBUS
Display with MODBUS
Display with local alarms and MODBUS
Smart sensor technology allows a user to install the transmitter or to replace a sensor within a few seconds – just plug the sensor into the transmitter. Adjustments are done directly at the transmitter using touch keys or with the RC2 remote control (one-person calibration).
The remote control display is always identical to that of the transmitter, making inspection, service, and calibration considerably simple. In addition, one remote control can be used for several transmitters, which helps cut costs and increase efficiency.
Certifications: ATEX-labelling: II 2G EEx em [ib] IIC T4 CE 0158.
EC-Type Examination Certificate: BVS 04 ATEX E 132 x.
Features: no safety barrier necessary.
Accessories information: MODBUS communication interface available for all transmitter configurations except Display/Alarm/Relay (004). Add “M” to the part number (e.g. 2811-17-001M) and add an additional fee, contact your representative in this case.
Ordering information: For all applications above –4 degrees farenheit, GfG recommends the standard NMH₃ sensor configuration. Please contact the manufacturer for proper selection of NH₃ sensors. For available accessories or additional information, contact your Avantor representative and see the attached datasheet and manual.