GMA 200 Series Fixed Controllers, Fixed Systems Gas Detection, GfG Instrumentation
Supplier: GFG INSTRUMENTATION, INC.
2200-004 2200-006 2200-008 2200-012 2200-016 2200-RT 2200-RTD 200MT-12 200MT-16 200MT-4 200MT-6 200MT-8
76334-840EA 2365.79 USD
76334-840 76334-842 76334-844 76334-846 76334-848 76334-856 76334-858 76334-920 76334-922 76334-944 76334-946 76334-948
GMA 200 Series Fixed Controllers, Fixed Systems Gas Detection, GfG Instrumentation
Gas Detection Systems
The GMA 200-MT can control up to 16 individual gas transmitters (sensors) and it is possible to extend the GMA 200 controller by 16 additional freely configurable relays.
  • Complete controller package available in 4 to 16 channel configurations
  • Up to 16 analog inputs or 16 digital inputs
  • Scalable system with fully programmable menu driven relays
  • NEMA 4X housing
  • Local display, stack light horn and strobe
  • Terminal blocks for easy termination
  • Power supply
Inputs:
16 Analog inputs: (4 to 20 mA or 0.2 to 1 mA); max. 50 Ohm input resistance.
Two digital inputs: acknowledgement of alarms can be freely configured.
2X RS485 BUS: for connection of external relay modules or digital transmitters in BUS wiring.
1X RS485 BUS: for digital transfer of measured and output data to a higher-level control centre or with master functionality of a GMA 200-MT for the connection of GMA 200-RT relay modules.
The clearly structured layout with graphical display enables the quick detection of hazardous situations. Currently measured values are displayed on the LCD graphical display.
Accessories information: The Stack Light adds an additional 150 mm (5.9") in height.
Ordering information: Sensors sold separately. For available sensors, relay module panels, and accessories or additional information, contact your Avantor representative and see the attached datasheets and manual.