Heavy Duty Epoxy Coated Alloy Clamp, Chemglass
Supplier: Chemglass
CG-2040-C-20
76333-686EA 68.52 USD
76333-686
Heavy Duty Epoxy Coated Alloy Clamp, Chemglass
Clamps General Purpose Clamps
Can be used to support stirrers, temperature monitors/condensers.
Heavy duty epoxy coated alloy clamp has high tensile strength and corrosion resistance that can be used to support stirrers, temperature monitors and condensers. Large hand knobs allow easy adjusment and secure clamping power. Useful wherever clamping at 90° is required.