Chem-Spin Scapers, Rotary Tools, Chemglass
Supplier: Chemglass
CG-1981-C3000 CG-1981-C5000 CG-1981-C100 CG-1981-C1000 CG-1981-C-24 CG-1981-C250 CG-1981-C-26 CG-1981-C-28 CG-1981-C-29
76333-390EA 108.51 USD
76333-390 76333-392 76333-638 76333-640 76333-642 76333-644 76333-646 76333-648 76333-650
Chem-Spin Scapers, Rotary Tools, Chemglass
Scrapers
Chem-spin scraper tools provide an easy method of removing the majority of desired, dried-on solids from the interior walls of round bottom flasks.
  • Stainless steel and PTFE for rapid sample removal
  • Choose from seven sizes- for 100 ml thru 5 L round bottom flasks
  • Laser etched as to the size and part number
  • Available individually, in tool kits and in motor kits
  • Fits outer joint size 24/40 or Larger
  • Patent pending design
Motor driven, the spinning chain rapidly and effectively brushes the surface as the loose solids collect on the bottom. Minimal physical effort is required. Tedious, time-consuming, manual scraping with a spatula is eliminated. Individual Rotary Scraper Tools are available to fit seven, popular, round bottom flask sizes: 100, 250 and 500 ml as well as 1, 2, 3 and 5 L . Tool Kits include selections of rotary scraper tools only.
Delivery information: Motor Kits include everything you need to get started: rotary scraper tools, glass bushing adapters, motors and two 12 V batteries.