Corepoint Scientific™ Premier™ Hydrocarbon Manual Defrost Solid Door Freezer, Horizon Scientific
Supplier: Horizon Scientific
NSPF141WWW/0MHC
76333-622EA 3519.47 USD
76333-622
Corepoint Scientific™ Premier™ Hydrocarbon Manual Defrost Solid Door Freezer, Horizon Scientific
Freezers Laboratory / General Purpose Freezers
These efficient manual defrost freezers provide premier cooling for laboratory settings. They offer storage solutions for a variety of scientific and clinical applications.
  • Microprocessor temperature control
  • Integrated temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
Units come equipped with forced draft circulation and precise temperature monitoring through microprocessor control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: *Select models ENERGY STAR certified: 76333-622
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.