CHEMRXNHUB™ Dual Support Stands, Benchtop Reactors, Chemglass
Supplier: Chemglass
CG-1949-X-20
76333-364EA 8028.72 USD
76333-364
CHEMRXNHUB™ Dual Support Stands, Benchtop Reactors, Chemglass
Supports Flask Supports
Supports 2 reactors side by side from 300 ml to 5 L. This unique stationary reactor stand is designed to fit into most benchtop hoods.
  • Stainless steel uprights and heavy duty PTFE coated aluminum fittings provide maximum strength and chemical durability
  • Supports two reactors side by side from 300 ml to 5 L
  • Removable polyethylene spill tray
  • Adjustable middle support bracket is vertically adjustable: It accepts our 60,100 and 150 mm S.S. support clamps
  • Adjustable top support bracket for each overhead motor is vertically adjustable and accommodates most brands of overhead stirrers
  • Mounting clamps accept overhead stirrers with a MAX support arm of ⁵/₈" diameter
  • Unique design allows easy removal/installation of glassware
  • Compact footprint
ChemRxnHub dual stand can hold two reactors from 300 ml to 5 L. The stand provides support for the reaction vessel(s), glassware, overhead stirring motor, and temperature monitor.
Delivery information: Stand includes removable spill tray.