VWR® PCR WorkUp Racks
Supplier: VWR International
5240-09-22AV1
76333-292CS 511.78 USD
76333-292
VWR® PCR WorkUp Racks
Racks Tube Racks
These racks accommodate 96-well PCR plates and 8 or 12 strip tubes.
  • Stackable
  • Lid locks into place
  • Stable work platform
Operating Temperature: –80 to 121 °C (maximum)
Delivery information: Supplied with one lid per rack.