External SmartChemical Reader, Mettler Toledo
Supplier: Mettler Toledo
30486743
76333-290EA 610 USD
76333-290
External SmartChemical Reader, Mettler Toledo
Titrators
External SmartChemical reader for titrator.
  • Error-free transfer from chemicals to titrator
  • Secure, traceable information flow ensures complete and correct reagent data
  • Efficiently reduce setup time with automated reagent data transfer
SmartChemicals titration chemicals and reagents automatically transfer their identifying data to the titrator. A simple contactless touch of reagent bottles to a dedicated SmartReader on select titrators instantly transfers all relevant information including the reagent name, lot, batch number, concentration, shelf life, open date, and expiration.
A quick scan eliminates all errors and extra time required for manual transcription.