Vacuum Gauge, Bargraph, 1000-1 Micron Complete with Thermocouple, Digivac
Supplier: Chemglass
AF-0302-10
76333-046EA 874.83 USD
76333-046
Vacuum Gauge, Bargraph, 1000-1 Micron Complete with Thermocouple, Digivac
Pumps Vacuum Gauges
Compact, low cost unit in a rugged metal case with solid state circuitry.
  • Display is easy to read, single point LED bar-graph with a single process set-point
The 4-Pin Varian style gauge tube is threaded with a ¹/₈" male NPT for use with the AF-0306 adapter. Supplied complete with a gauge tube having a 10 foot cord.