VWR® Microcentrifuge Tubes, Capless
Supplier: VWR International
1270-00-201AV
76332-078CS 905.9 USD
76332-078
VWR® Microcentrifuge Tubes, Capless
Tubes Microcentrifuge Tubes
These microcentrifuge tubes are available without caps. Best for use in fraction collection and whenever storage of samples is not necessary.
  • Rated 20000 ×g
  • Graduated every 100 µl
  • Polypropylene
Capless tubes are graduated and are available in natural only.