Corning® Matrigel® Matrix for Organoid Culture
Supplier: Corning
356255
76332-052EA 468.9 USD
76332-052
Corning® Matrigel® Matrix for Organoid Culture
Cell Culture Supplements and Additives
Corning Matrigel Matrix for Organoid Culture is a solubilized basement membrane preparation that has been optimized for organoid culture.
  • Quality tested for ability to form and maintain stable 3D domes/droplets on multi-well plates
  • Elastic Modulus (matrix stiffness) measured using a rheometer
  • Protein concentrations determined by Lowry method
  • Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
  • Extensive PCR testing is performed to screen for a number of pathogens
  • Tested and found negative for bacteria, fungi, and mycoplasma
  • Endotoxin units measured by Limulus Amoebocyte Lysate assay
Verified to support both mouse and human organoid growth and differentiation from healthy and/or diseased tissue, each lot has a specific elastic modulus value indicating the stiffness of the gel formed and is qualified to form stable 3D domes commonly used in organoid culture protocols. Available in 10 ml LDEV-free, Phenol Red-free format.
Corning Matrigel Matrix for Organoid culture is extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.