Metal Lock Box
Supplier: Therapak, LLC
365172
76330-068EA 187.63 USD
76330-068
Metal Lock Box
Boxes Transport Boxes
Each box is made with cold rolled steel for durability and powder-coated white for protection. It is insulated on the inside with foam to assist with preserving specimen integrity. A standard-keyed lock with 2 keys is provided with each unit.
  • Durable and made in the USA
  • Permanently attached straps
  • Customization available
Standard color: White
Standard printing: Reflex blue text, red biohazard symbol on front side only
Customization: Any deviation from the standard configuration, which may include changes in color, size, design features, closure and print.
Minimum quantity for custom print is 150 units.