Loopster Digital Rotating Shaker, IKA
Supplier: IKA Works
0004016000
76329-832EA 1796.63 USD
76329-832
Loopster Digital Rotating Shaker, IKA
Shakers and Mixers Rotating/Rocking Shakers
Digital rotator for smooth but effective mixing with adjustable speed. Ideal for biological samples, particularly blood samples.
  • Usable at 4 to 50 °C in incubators
  • Freely adjustable rotary angle
  • Suitable for continuous operation
  • Error code display
  • Easy operation by foil keypad
  • Timer: Countdown, adjustable from 1 second to 5999 minutes
Suitable for separating and mixing processes of powdered and liquid samples in Eppendorf tubes and Greiner tubes up to 50 ml. The easy removable attachments allow simple and quick cleaning of the unit of spilled samples.
Certifications: IP 21 protection class according to DIN EN 60529.