Magnetic Stirrer, KMO 3 basic, IKA
Supplier: IKA Works
0020004631
76329-828EA 1027.92 USD
76329-828
Magnetic Stirrer, KMO 3 basic, IKA
Stirrers Magnetic Stirrers
Compact magnetic stirrer with high quality, fast-response display and hardened glass work surface with a brilliant, clear white central area to easily view color changes.
  • Torque display
  • Right and left rotation
  • Illuminated, flat stirring plate
  • Key lock button
  • Integrated USB interface (control via labworldsoft and use of Firmware Update Tool)
  • Recessed thread to accommodate support rod
The permissible relative humidity is 80%. It has automatic reverse rotation, set-up plate illuminated, LED display and permissible ambient temperature of 5 to 40 °C.
Certifications: IP protection class according to DIN EN 60529: IP 42.