Vortex Shaker, MS 3 control, IKA
Supplier: IKA Works
0020016017
76329-824EA 1232.32 USD
76329-824
Vortex Shaker, MS 3 control, IKA
Shakers and Mixers Vortex Mixers
Compact, universal small shaker suitable for shaking tasks with all small vessels and microtiter plates.
  • Wide range of attachments
  • Attachment detection
  • Timer with countdown function
  • Stable in all speed ranges
  • Sturdy zinc die cast casing
  • USB connection for remote control
  • Continuous or touch operation (with standard attachment)
Two operating modes, Mode A (safe mode with attachment detection) - The maximum speed of 3000 rpm is only reached with the standard attachment in touch mode. When using other attachments the speed is limited to 1300 rpm.
Mode B (without attachment detection) - A speed of 3000 rpm is possible with all attachments, depending on the application (see accessory list in the manual).
RS232 interface with LCD timer and speed display. Permissible relative humidity is 80%.
Certifications: Protection class according to DIN EN 60529: IP 21