Mini G Centrifuge, IKA
Supplier: IKA Works
0003958000
76329-822EA 781.16 USD
76329-822
Mini G Centrifuge, IKA
Centrifuges Benchtop Centrifuges
Mini-centrifuge for all applications which do not require high speeds, such as micro-filtrations and cell separation
  • Fixed speed of 6000 rpm
  • Integrated quick stop
  • Fast startup of speed
  • Compact design and quiet operation
  • Rotor exchangeable without tools
  • Easy operation with touch keypad
  • Protective cover
The transparent cover allows constant observation of centrifuge contents. The mini-centrifuge can be used for both PCR vessels and PCR strips. As a safety feature, the centrifuge will run only when the lid is closed. The lid opens easily with the press of a button.
Speed deviation of about 5%.
Delivery information: It includes rotor 8×1.5/2 ml, rotor 4×strip 8×0.2ml, adapter set 0.5 ml×8, adapter set 0.4 ml×8 and adapter set 0.2 ml×8.