Calculi Strainers for 24-Hour Urine Containers, Medegen Medical Products
Supplier: Medegen Medical Products, LLC
4712
76329-592CS 174.46 USD
76329-592
Calculi Strainers for 24-Hour Urine Containers, Medegen Medical Products
Cell Strainers
Calculi strainers provide quick and easy mesh filtering for kidney stones or other foreign matter.
  • Designed to remain stable a top the 24-hour container throughout the entire filtering process
  • Built-in ridge serves as a handle for simple application and removal
  • Latex-Free
Strainer fits securely onto both the 76329-584 and 76329-586 24-hour specimen container.