Easy Bev Benchtop Refractometer, Mettler-Toledo
Supplier: Mettler Toledo
30468674
76329-512EA 8265.93 USD
76329-512
Easy Bev Benchtop Refractometer, Mettler-Toledo
Refractometers Benchtop Refractometers
Easy Bev measures refractive index and Brix with automatic temperature control. It offers a ideal solution for beverage manufacturers with the smart vertical design.
This intuitive instrument allows simple operature with immediate QC results.