Easy Brix Benchtop Refractometer, Mettler-Toledo
Supplier: Mettler Toledo
30468673
76329-510EA 6890 USD
76329-510
Easy Brix Benchtop Refractometer, Mettler-Toledo
Refractometers Benchtop Refractometers
The Easy Brix refractometer offers simple and fast refractive index and Brix measurements with automatic temperature control.
  • Compact and robust design
Suitable for use in the laboratory or near the production area.