Easy R40 Benchtop Refractometer, Mettler-Toledo
Supplier: Mettler Toledo
30468672
76329-508EA 10100 USD
76329-508
Easy R40 Benchtop Refractometer, Mettler-Toledo
Refractometers Benchtop Refractometers
The Easy R40 refractometer offers fast refractive index and Brix measurements with automatic temperature control.
  • Compact and robust design
  • Suitable for use in the laboratory or near the production area
Using only a small sample volume, the Easy R40 provide results within seconds.