Stuart® 8500 rpm SCF4 Microcentrifuge, Cole Parmer
Supplier: Antyila Scientific
17414-70
76327-228EA 320.75 USD
76327-228
Stuart® 8500 rpm SCF4 Microcentrifuge, Cole Parmer
Centrifuges Microcentrifuges
The Stuart 8500 microcentrifuge provides high speed centrifugation with enhanced safety features.
  • Personal size for quick spin downs
  • Safely spin samples with dual locking lid
  • Quickly change rotors with quick-release adapter, no tools required
  • Built-in recessed rotor bowl contains spills for easy clean-up
  • Two year warranty
The high speed 8500 rpm and 3884 RCF rating allows users to spin samples rapidly or be used in applications where greater g-force is required.
Certifications: CE approved.
Delivery information: Includes fixed-angle rotor for 8×1.5 to 2.0 ml tubes, rotor for 32×0.2 ml strip tubes, 8×0.5 ml adapter inserts and detachable power cords for US, UK and EU plugs.