Elma Lab Clean A20Sf Pipette Cleaning Solution
Supplier: Elma
8000057
76326-584EA 201.46 USD
76326-584
Elma Lab Clean A20Sf Pipette Cleaning Solution
Cleaning Agents Detergents and Surface Cleansers
Elma Lab Clean A20SF solution cleans pipettes and volumetric ware in ultrasonic cleaners and spray washers. Cleaned glass surfaces are free of surfactants and allow undisturbed draining of liquids.
  • Pipettes cleaned with A20SF guarantee volumetric precision
  • A20SF concentrate is diluted to 1% before use in ultrasonic cleaner
Mildly alkaline, surfactant-free, Elma Lab Clean A20SF is used in ultrasonic cleaning baths and pipette rinsing units. The solution removes a broad range of contaminants including light grease, lime soap residues, fingerprints, and dust.
Elma Lab Clean A20SF cleans volumetric glassware such as calibrated pipettes, burettes, and graduated cylinders, as well as volumetric instruments made of stainless steel, ceramics and plastic materials. Not for aluminum, magnesium and alloys of light-metals.
The concentrated solution should be diluted with water to 1% for use in ultrasonic cleaners, 0.5% in spray washers, and > 2% for soaking.
Safe and cost effective Elma cleaning solutions prolong the lifetime of your ultrasonic bath.