SP Bel-Art Poxygrid® Poxygrid® Hybridization Bottle Rack, SP Scienceware, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F16964-0006
76326-304EA 137.51 USD
76326-304
SP Bel-Art Poxygrid® Poxygrid® Hybridization Bottle Rack, SP Scienceware, Bel-Art Products, a part of SP
Racks Bottle Racks
Heavy-duty rack protects your expensive bottles keeping them from rolling, tipping and striking against one another.
  • Epoxy-coated steel rods provide durability and easy cleaning
  • Steam autoclavable at 121 °C (250 °F)
  • Holes are 42 mm square shaped for holding 40 mm diameter bottles
Holds 150 and 300 mm long bottles with ample space in between to allow for the screw cap diameter and provide convenient handling.