VWR® Traceable® Indoor/Outdoor Digital Thermometer with Giant Dual-Display and Calibration
Supplier: VWR International
76326-136
76326-136EA 87.46 USD
76326-136
VWR® Traceable® Indoor/Outdoor Digital Thermometer with Giant Dual-Display and Calibration
Thermometers Digital Thermometers
Jumbo digital display is easy to read from distances.
  • Get accurate, reliable measurements with included NIST-traceable calibration
  • F/°C selectable
  • Hi/Lo alarm
NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.
Certifications: NIST-Traceable
Delivery information: Includes a probe, one AAA battery, and NIST-traceable calibration certificate with data.