Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
The Color prestained protein standard, broad range is a mixture of highly pure, recombinant, prestained proteins, covalently coupled with a blue chromophore, and two reference bands (one orange and one green at 72 kDa and 26 kDa, respectively), that resolves into 11 sharp bands when electrophoresed.
- Allows approximate molecular weight determination when performing SDS-PAGE analysis
- Applications include verification of Western transfer efficiency and fluorescent imaging of SDS-PAGE
- Direct loading, additional loading buffer and heat incubation not required
- Recombinant proteins with no detectable protease contaminating activities
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...