Tte-UvrD Helicase
Supplier: New England Biolabs (NEB)
M1202S
76326-038EA 84 USD
76326-038
Tte-UvrD Helicase
Enzymes
Tte UvrD Helicase is a repair helicase capable of unwinding double-stranded DNA, without a requirement for a specific flap or overhang structure, from the thermophilic organism Thermoanaerobacter tengcongensis.
  • Unwinds double-stranded DNA
  • Thermostable to 65 °C
  • Reduces non-specific product formation in isothermal amplification (LAMP)
It is active on a wide range of DNA substrates and, along with its thermostability (active to 70 °C), Tte UvrD Helicase has been demonstrated to be a useful additive for improving specificity of isothermal amplification reactions, particularly in conjunction with the WarmStart LAMP kit.