Anti-Mouse CyDye™ 800 NIR-labelled secondary antibody

Supplier: Cytiva
29-3607-88 29-3607-89
76325-084EA 140.45 USD
76325-084 76325-086
Anti-Mouse CyDye™ 800 NIR-labelled secondary antibody
Antibodies
Amersham CyDye NIR-labeled secondary antibodies enable quantitation of Western blots with a strong signal-to-noise ratio. Mulitplexed experiments allow for the detection of two proteins on a single blot. The use of total lane signals to normalize target protein data yield robust and reliable results.

  • Easy handling and a strong signal-to-noise ratio yield reproducible results
  • Multiplexing analysis of more than one target protein on a single blot increases process efficiency
  • Total protein normalization (TPN) generates reliable and accurate quantitation
  • Stable signals allow membranes to be stored and rescanned if needed for future studies

Amersham CyDye 700 and 800 are secondary antibodies labeled with NIR fluorophores. They emit light at wavelengths of 700 nm and 800 nm. Multiplexed experiments can be performed by using both of these secondary antibodies along with a pair of appropriate rabbit or mouse primary antibodies.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: CyDye™ 800
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR