Polyribonucleotide poly(I)·poly(C), Cytiva
Supplier: Cytiva
27-4732-01
76325-074EA 3298.04 USD
76325-074
Polyribonucleotide poly(I)·poly(C), Cytiva
Cell Culture Supplements and Additives
The double-stranded polyribonucleotide poly(I)·poly(C) induces interferon in rabbit cells, and there has been interest in its effectiveness for inducing human interferon in vitro.