HiScreen™ Capto PlasmidSelect, Columns, Cytiva
Supplier: Cytiva
29-2017-90
76325-064EA 388.97 USD
76325-064
HiScreen™ Capto PlasmidSelect, Columns, Cytiva
Chromatography Columns
Columns packed with Capto PlasmidSelect are suitable for purification of supercoiled DNA to high quality for gene therapy and DNA vaccine applications. Capto PlasmidSelect is a chromatography resin that allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.
  • High-flow purification of supercoiled DNA from research scale to cGMP production
  • Flexible process design due to a large operational window of flow velocities and bed heights
  • Higher throughput for improved productivity and process economy
  • Hydrophilic properties of base matrix prevent non-specific binding
  • BioProcess resin supported for industrial applications
  • Convenient small-scale purification, process development, and scale-up using prepacked HiScreen™ columns
Capto PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production.